siRNA Details
| Gene Symbol: Mst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024352.1 | NP_077328.1 | 8 | 991 | ||
| XM_006243706.1 | XP_006243768.1 | 8 | 1102 | |||
| GenBank mRNA | BC089796.1 | - | 8 | 995 | 7322260 | |
| X95096.1 | - | 8 | 991 | |||

Mst1
Genomic Map