siRNA Details
Gene Symbol: Exoc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139043.1 | NP_620612.1 | 1 | 1551 | ||
GenBank mRNA | AF032669.1 | - | 1 | 1551 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Exoc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139043.1 | NP_620612.1 | 1 | 1551 | ||
GenBank mRNA | AF032669.1 | - | 1 | 1551 |