siRNA Details
| Gene Symbol: Magi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139084.2 | NP_620784.2 | 14 | 2499 | ||
| XM_006233058.2 | XP_006233120.1 | 14 | 2487 | |||
| XM_008761367.1 | XP_008759589.1 | 14 | 2487 | |||
| GenBank mRNA | AF255614.1 | - | 14 | 2508 | ||

Magi3
Genomic Map