siRNA Details
| Gene Symbol: Mme | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012608.2 | NP_036740.1 | 23 | 2317 | ||
| XM_006232437.2 | XP_006232499.1 | 23 | 2353 | |||
| GenBank mRNA | BC085753.1 | - | 23 | 2317 | 7110842 | |
| M15944.1 | - | 22 | 2267 | |||

Mme
Genomic Map