siRNA Details
| Gene Symbol: Cgref1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139087.2 | NP_620787.2 | 6 | 763 | ||
| GenBank mRNA | FQ220335.1 | - | 6 | 693 | ||
| U66470.1 | - | 6 | 684 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cgref1| Gene Symbol: Cgref1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139087.2 | NP_620787.2 | 6 | 763 | ||
| GenBank mRNA | FQ220335.1 | - | 6 | 693 | ||
| U66470.1 | - | 6 | 684 | |||