siRNA Details
| Gene Symbol: Scaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139094.1 | NP_620794.1 | 15 | 1972 | ||
| XM_006227858.2 | XP_006227920.1 | 16 | 1899 | |||
| GenBank mRNA | U49055.1 | - | 15 | 1972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scaf8| Gene Symbol: Scaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139094.1 | NP_620794.1 | 15 | 1972 | ||
| XM_006227858.2 | XP_006227920.1 | 16 | 1899 | |||
| GenBank mRNA | U49055.1 | - | 15 | 1972 | ||