siRNA Details
| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139096.1 | NP_620796.1 | 6 | 1493 | ||
| GenBank mRNA | AF065438.1 | - | 6 | 1493 | ||
| AY552591.1 | - | 5 | 1355 | |||
| BC081724.1 | - | 6 | 1466 | 7123606 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgals3bp| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139096.1 | NP_620796.1 | 6 | 1493 | ||
| GenBank mRNA | AF065438.1 | - | 6 | 1493 | ||
| AY552591.1 | - | 5 | 1355 | |||
| BC081724.1 | - | 6 | 1466 | 7123606 | ||