siRNA Details
| Gene Symbol: Scn3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139097.3 | NP_620797.1 | 5 | 920 | ||
| XM_006243102.1 | XP_006243164.1 | 5 | 1002 | |||
| XM_006243103.1 | XP_006243165.1 | 5 | 1002 | |||
| GenBank mRNA | AF378093.1 | - | 5 | 619 | ||
| AJ243395.2 | - | 5 | 905 | |||
| BC070899.1 | - | 5 | 920 | 7100476 | ||
| FQ211809.1 | - | 2 | 214 | |||

Scn3b
Genomic Map