siRNA Details
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270762.1 | NP_001257691.1 | 4, 5 | 538 | ||
NM_001270763.1 | NP_001257692.1 | 4, 5 | 544 | |||
NM_139105.3 | NP_620805.2 | 4, 5 | 553 | |||
XM_008759970.1 | XP_008758192.1 | 4, 5 | 1102 | |||
GenBank mRNA | AB296102.1 | - | 3, 4 | 420 | ||
BC070501.1 | - | 4, 5 | 460 | 7096144 | ||
FQ233708.1 | - | 4, 5 | 478 | |||
FQ234889.1 | - | 4, 5 | 477 | |||
X62528.1 | - | 4, 5 | 538 |
