siRNA Details
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270762.1 | NP_001257691.1 | 9 | 1317 | ||
NM_001270763.1 | NP_001257692.1 | 9 | 1323 | |||
NM_139105.3 | NP_620805.2 | 9 | 1332 | |||
XM_008759970.1 | XP_008758192.1 | 9 | 1881 | |||
GenBank mRNA | AB296102.1 | - | 8 | 1199 | ||
BC070501.1 | - | 9 | 1239 | 7096144 | ||
FQ233708.1 | - | 9 | 1257 | |||
FQ234889.1 | - | 9 | 1259 | |||
X62528.1 | - | 9 | 1317 |
