siRNA Details
| Gene Symbol: Ngfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012610.2 | NP_036742.2 | 3 | 527 | ||
| GenBank mRNA | X05137.1 | - | 3 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ngfr| Gene Symbol: Ngfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012610.2 | NP_036742.2 | 3 | 527 | ||
| GenBank mRNA | X05137.1 | - | 3 | 527 | ||