siRNA Details
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139109.2 | NP_620809.1 | 16 | 2160 | ||
GenBank mRNA | AF277899.1 | - | 16 | 2163 | ||
BC081723.1 | - | 16 | 2154 | 7108549 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139109.2 | NP_620809.1 | 16 | 2160 | ||
GenBank mRNA | AF277899.1 | - | 16 | 2163 | ||
BC081723.1 | - | 16 | 2154 | 7108549 |