siRNA Details
Gene Symbol: Nos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021838.2 | NP_068610.1 | 23 | 3094 | ||
XM_006235872.1 | XP_006235934.1 | 23 | 3123 | |||
GenBank mRNA | AB176831.1 | - | 23 | 3094 | ||
AF093837.1 | - | 1 | 46 | |||
AJ011116.1 | - | 3 | 240 | |||
AJ249546.1 | - | 6 | 826 | |||
AY695391.1 | - | 24 | 3028 | |||
U18336.1 | - | 1 | 38 |
