siRNA Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139189.2 | NP_631928.2 | 9 | 874 | ||
| GenBank mRNA | AY093598.1 | - | 9 | 786 | ||
| BC078723.1 | - | 9 | 874 | 7123105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmbrd1| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139189.2 | NP_631928.2 | 9 | 874 | ||
| GenBank mRNA | AY093598.1 | - | 9 | 786 | ||
| BC078723.1 | - | 9 | 874 | 7123105 | ||