siRNA Details
Gene Symbol: Calcoco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139190.1 | NP_631929.1 | 935 | |||
GenBank mRNA | AY078385.1 | - | 935 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Calcoco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139190.1 | NP_631929.1 | 935 | |||
GenBank mRNA | AY078385.1 | - | 935 |