siRNA Details
| Gene Symbol: Foxi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341949.4 | XP_341950.2 | 2 | 1778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxi2| Gene Symbol: Foxi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341949.4 | XP_341950.2 | 2 | 1778 | ||