siRNA Details
| Gene Symbol: Akap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037093.2 | NP_001032170.2 | 42 | 10583 | ||
| XM_006236023.2 | XP_006236085.1 | 44 | 10704 | |||
| XM_006236025.2 | XP_006236087.1 | 43 | 10713 | |||
| XM_008762685.1 | XP_008760907.1 | 45 | 10818 | |||
| XM_008762686.1 | XP_008760908.1 | 44 | 10779 | |||
| GenBank mRNA | BC098762.1 | - | 4 | 432 | ||
| DQ228948.2 | - | 13 | 2796 | |||

Akap9
Genomic Map