siRNA Details
| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100823.1 | NP_001094293.1 | 7, 8 | 1402 | ||
| GenBank mRNA | AF130121.1 | - | 7, 8 | 1165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr44| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100823.1 | NP_001094293.1 | 7, 8 | 1402 | ||
| GenBank mRNA | AF130121.1 | - | 7, 8 | 1165 | ||