siRNA Details
| Gene Symbol: Kcnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139216.1 | NP_631962.1 | 2 | 833 | ||
| NM_139217.1 | NP_631963.1 | 2 | 790 | |||
| XM_006241327.2 | XP_006241389.1 | 3 | 1042 | |||
| GenBank mRNA | M34052.1 | - | 2 | 833 | ||
| M59211.1 | - | 2 | 790 | |||
| M84202.1 | - | 3 | 847 | |||
| M84203.1 | - | 2 | 833 | |||
| X62839.1 | - | 3 | 944 | |||

Kcnc2
Genomic Map