siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012619.2 | NP_036751.2 | 5 | 567 | ||
| XM_008765215.1 | XP_008763437.1 | 3 | 507 | |||
| GenBank mRNA | BC078881.1 | - | 5 | 567 | 7131414 | |
| FQ219680.1 | - | 5 | 597 | |||
| M12337.1 | - | 5 | 552 | |||

Pah
Genomic Map