siRNA Details
| Gene Symbol: Serpine1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012620.1 | NP_036752.1 | 4 | 804 | ||
| GenBank mRNA | M24067.1 | - | 4 | 804 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpine1| Gene Symbol: Serpine1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012620.1 | NP_036752.1 | 4 | 804 | ||
| GenBank mRNA | M24067.1 | - | 4 | 804 | ||