siRNA Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198050.1 | NP_932167.1 | 4 | 1116 | ||
| XM_006253556.2 | XP_006253618.1 | 4 | 1260 | |||
| XM_006253557.2 | XP_006253619.1 | 4 | 1260 | |||
| XM_008771424.1 | XP_008769646.1 | 4 | 1284 | |||

Kif27
Genomic Map