siRNA Details
| Gene Symbol: Lrit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139331.2 | NP_647547.2 | 3 | 696 | ||
| GenBank mRNA | AB028461.1 | - | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrit1| Gene Symbol: Lrit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139331.2 | NP_647547.2 | 3 | 696 | ||
| GenBank mRNA | AB028461.1 | - | 696 | |||