siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139336.1 | NP_647552.1 | 3, 4 | 218 | ||
| GenBank mRNA | AF482705.1 | - | 3, 4 | 218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uxs1| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139336.1 | NP_647552.1 | 3, 4 | 218 | ||
| GenBank mRNA | AF482705.1 | - | 3, 4 | 218 | ||