siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139336.1 | NP_647552.1 | 15 | 1203 | ||
| XM_006244296.2 | XP_006244358.1 | 15 | 1261 | |||
| XM_008766751.1 | XP_008764973.1 | 16 | 1719 | |||
| GenBank mRNA | AF482705.1 | - | 15 | 1203 | ||
| BC086988.1 | - | 15 | 1214 | 7192831 | ||

Uxs1
Genomic Map