siRNA Details
Gene Symbol: Plekha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763357.1 | XP_008761579.1 | 9 | 816 | ||
XM_008763358.1 | XP_008761580.1 | 9 | 816 | |||
XM_008763359.1 | XP_008761581.1 | 9 | 816 | |||
XM_008763360.1 | XP_008761582.1 | 9 | 816 | |||
XM_008763361.1 | XP_008761583.1 | 7 | 1507 | |||
XM_008763362.1 | XP_008761584.1 | 9 | 816 | |||
XM_008763363.1 | XP_008761585.1 | 2 | 644 | |||
XM_008763364.1 | XP_008761586.1 | 9 | 816 | |||
GenBank mRNA | AF468695.1 | - | 7 | 565 |
