siRNA Details
| Gene Symbol: Slc15a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139341.1 | NP_647557.1 | 1 | 560 | ||
| GenBank mRNA | AB026665.1 | - | 1 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc15a3| Gene Symbol: Slc15a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139341.1 | NP_647557.1 | 1 | 560 | ||
| GenBank mRNA | AB026665.1 | - | 1 | 560 | ||