siRNA Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139342.1 | NP_647558.1 | 6 | 1045 | ||
| XM_006251928.2 | XP_006251990.1 | 5 | 1897 | |||
| XM_008770661.1 | XP_008768883.1 | 4 | 858 | |||
| XM_008770662.1 | XP_008768884.1 | 5 | 1229 | |||
| GenBank mRNA | AF036537.2 | - | 6 | 1045 | ||
| BC158580.1 | - | 4 | 612 | 7378607 | ||

Ripk3
Genomic Map