siRNA Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144737.2 | NP_653338.2 | 4 | 477 | ||
| XM_008769604.1 | XP_008767826.1 | 3 | 741 | |||
| XM_008769605.1 | XP_008767827.1 | 4 | 551 | |||
| GenBank mRNA | AF458414.1 | - | 4 | 444 | ||
| BC070904.1 | - | 4 | 477 | 7100688 | ||
| CK365136.1 | - | 4 | 412 | |||

Fmo2
Genomic Map