siRNA Details
| Gene Symbol: Fmo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144561.2 | NP_653147.1 | 10 | 1885 | ||
| NM_144562.1 | NP_653148.1 | 8 | 1490 | |||
| XM_006250144.2 | XP_006250206.1 | 10 | 1820 | |||
| GenBank mRNA | AF458416.1 | - | 10 | 1886 | ||
| AF458417.1 | - | 8 | 1490 | |||
| BC081721.1 | - | 10 | 1875 | 7107835 | ||

Fmo4
Genomic Map