siRNA Details
| Gene Symbol: Arhgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144740.3 | NP_653341.2 | 21 | 2746 | ||
| XM_006229545.2 | XP_006229607.1 | 21 | 3506 | |||
| XM_006229546.2 | XP_006229608.1 | 20 | 3443 | |||
| XM_006229547.2 | XP_006229609.1 | 20 | 3404 | |||
| XM_006229548.2 | XP_006229610.1 | 20 | 3272 | |||
| GenBank mRNA | AF401635.1 | - | 21 | 2756 | ||
| AY742900.1 | - | 2759 | ||||

Arhgap4
Genomic Map