siRNA Details
| Gene Symbol: Ppp2r2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144746.2 | NP_653347.2 | 5, 6 | 583 | ||
| GenBank mRNA | AF180350.1 | - | 5, 6 | 587 | ||
| BC081720.1 | - | 5, 6 | 583 | 7112646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r2d| Gene Symbol: Ppp2r2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144746.2 | NP_653347.2 | 5, 6 | 583 | ||
| GenBank mRNA | AF180350.1 | - | 5, 6 | 587 | ||
| BC081720.1 | - | 5, 6 | 583 | 7112646 | ||