siRNA Details
| Gene Symbol: Oas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144752.2 | NP_653353.2 | 4 | 826 | ||
| GenBank mRNA | AF068268.1 | - | 838 | |||
| BC091121.1 | - | 4 | 826 | 7383389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oas1b| Gene Symbol: Oas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144752.2 | NP_653353.2 | 4 | 826 | ||
| GenBank mRNA | AF068268.1 | - | 838 | |||
| BC091121.1 | - | 4 | 826 | 7383389 | ||