siRNA Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017031.2 | NP_058727.2 | 12 | 1864 | ||
| GenBank mRNA | AF202732.1 | - | 1722 | |||
| AF202733.1 | - | 12 | 1428 | |||
| J04563.1 | - | 9 | 876 | |||
| L27058.1 | - | 7 | 1423 | |||
| M25350.1 | - | 5 | 489 | |||
| M28413.1 | - | 5 | 489 | |||
| U95748.1 | - | 12 | 1864 | |||

Pde4b
Genomic Map