siRNA Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017031.2 | NP_058727.2 | 1 | 555 | ||
| GenBank mRNA | U95748.1 | - | 1 | 555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde4b| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017031.2 | NP_058727.2 | 1 | 555 | ||
| GenBank mRNA | U95748.1 | - | 1 | 555 | ||