siRNA Details
| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144756.2 | NP_653357.1 | 10, 11 | 809 | ||
| GenBank mRNA | AF044201.1 | - | 10, 11 | 758 | ||
| BC087606.1 | - | 10, 11 | 809 | 7314068 | ||
| Gene Symbol: LOC102551901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242293.2 | XP_006242355.1 | 10, 11 | 1035 | ||

Faim2
Genomic Map