siRNA Details
| Gene Symbol: Optn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145081.3 | NP_659549.2 | 6 | 856 | ||
| GenBank mRNA | AB069907.3 | - | 6 | 856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Optn| Gene Symbol: Optn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145081.3 | NP_659549.2 | 6 | 856 | ||
| GenBank mRNA | AB069907.3 | - | 6 | 856 | ||