siRNA Details
| Gene Symbol: Pdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145091.4 | NP_659559.2 | 2 | 1050 | ||
| XM_006255048.2 | XP_006255110.1 | 2 | 1095 | |||
| GenBank mRNA | AF062741.1 | - | 2 | 950 | ||
| BC072485.1 | - | 1050 | 7105072 | |||

Pdp2
Genomic Map