siRNA Details
| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF061957.1 | - | 7 | 1127 | ||
| AJ007632.1 | - | 7 | 1051 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh8| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF061957.1 | - | 7 | 1127 | ||
| AJ007632.1 | - | 7 | 1051 | |||