siRNA Details
Gene Symbol: Zdhhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145096.2 | NP_659564.2 | 4 | 527 | ||
XM_008771288.1 | XP_008769510.1 | 4 | 557 | |||
GenBank mRNA | AB217870.1 | - | 4 | 326 | ||
AF228917.1 | - | 4 | 516 | |||
AY886521.1 | - | 4 | 527 |
