siRNA Details
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017033.2 | NP_058729.2 | 9 | 1437 | ||
XM_006238442.2 | XP_006238504.1 | 10 | 1628 | |||
XM_008763853.1 | XP_008762075.1 | 9 | 1980 | |||
GenBank mRNA | BC099807.1 | - | 9 | 1437 | 7937943 | |
BC128703.1 | - | 9 | 1442 | |||
FQ220427.1 | - | 2 | 227 | |||
L11694.1 | - | 9 | 1420 |
