siRNA Details
| Gene Symbol: Serpina1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022519.2 | NP_071964.2 | 3 | 369 | ||
| XM_006240456.1 | XP_006240518.1 | 2 | 226 | |||
| GenBank mRNA | BC078824.1 | - | 3 | 369 | 7131535 | |
| D00675.1 | - | 2 | 172 | |||
| FQ218411.1 | - | 2 | 301 | |||
| FQ219741.1 | - | 2 | 163 | |||
| M32247.1 | - | 1 | 109 | |||

Serpina1
Genomic Map