siRNA Details
| Gene Symbol: Pklr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012624.3 | NP_036756.3 | 11 | 1569 | ||
| XM_006232593.2 | XP_006232655.1 | 11 | 1700 | |||
| GenBank mRNA | BC161827.1 | - | 10 | 1559 | 7127842 | |
| FQ211172.1 | - | 5 | 997 | |||
| M11709.1 | - | 11 | 1570 | |||

Pklr
Genomic Map