siRNA Details
| Gene Symbol: Pklr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012624.3 | NP_036756.3 | 9 | 1279 | ||
| XM_006232593.2 | XP_006232655.1 | 9 | 1410 | |||
| GenBank mRNA | BC161827.1 | - | 8 | 1269 | 7127842 | |
| FQ211172.1 | - | 3 | 707 | |||
| M11709.1 | - | 9 | 1280 | |||

Pklr
Genomic Map