siRNA Details
| Gene Symbol: Prl6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022176.2 | NP_071512.2 | 4 | 647 | ||
| GenBank mRNA | BC158563.1 | - | 4 | 647 | 7443132 | |
| FQ219971.1 | - | 4 | 663 | |||
| FQ221549.1 | - | 4 | 664 | |||
| M31155.1 | - | 4 | 606 | |||

Prl6a1
Genomic Map