siRNA Details
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270619.1 | NP_001257548.1 | 2 | 1235 | ||
NM_001270620.1 | NP_001257549.1 | 3 | 1709 | |||
NM_033096.2 | NP_149087.1 | 2 | 1235 | |||
XM_008764454.1 | XP_008762676.1 | 1 | 846 | |||
GenBank mRNA | AJ271834.1 | - | 2 | 1228 | ||
AJ271837.1 | - | 2 | 1228 | |||
BC061986.1 | - | 2 | 1190 | 5599216 | ||
BC081762.1 | - | 3 | 1709 | 7113731 | ||
FM108050.1 | - | 1 | 390 | |||
S90449.1 | - | 2 | 1521 |
