siRNA Details
| Gene Symbol: Ppm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270619.1 | NP_001257548.1 | 3 | 1310 | ||
| NM_001270620.1 | NP_001257549.1 | 4 | 1784 | |||
| NM_033096.2 | NP_149087.1 | 3 | 1310 | |||
| XM_008764454.1 | XP_008762676.1 | 2 | 921 | |||
| GenBank mRNA | AJ271834.1 | - | 3 | 1303 | ||
| AJ271837.1 | - | 3 | 1303 | |||
| BC061986.1 | - | 3 | 1265 | 5599216 | ||
| BC081762.1 | - | 4 | 1784 | 7113731 | ||
| FM075941.1 | - | 1 | 50 | |||
| FM108050.1 | - | 2 | 465 | |||
| S90449.1 | - | 3 | 1596 | |||

Ppm1b
Genomic Map