siRNA Details
| Gene Symbol: Ppm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270619.1 | NP_001257548.1 | 2 | 835 | ||
| NM_001270620.1 | NP_001257549.1 | 3 | 1309 | |||
| NM_033096.2 | NP_149087.1 | 2 | 835 | |||
| XM_008764454.1 | XP_008762676.1 | 1 | 446 | |||
| GenBank mRNA | AJ271834.1 | - | 2 | 828 | ||
| AJ271837.1 | - | 2 | 828 | |||
| BC061986.1 | - | 2 | 790 | 5599216 | ||
| BC081762.1 | - | 3 | 1309 | 7113731 | ||
| S90449.1 | - | 2 | 1121 | |||

Ppm1b
Genomic Map