siRNA Details
| Gene Symbol: Ppp1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031527.1 | NP_113715.1 | 5 | 623 | ||
| GenBank mRNA | BC070517.1 | - | 5 | 603 | 7104674 | |
| D00859.1 | - | 5 | 623 | |||
| D90163.1 | - | 5 | 607 | |||
| FQ224860.1 | - | 5 | 613 | |||
| FQ231613.1 | - | 5 | 632 | |||
| FQ232496.1 | - | 5 | 631 | |||
| FQ233040.1 | - | 5 | 623 | |||
| FQ235125.1 | - | 5 | 623 | |||

Ppp1ca
Genomic Map