siRNA Details
| Gene Symbol: Ppp3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017041.1 | NP_058737.1 | 6 | 865 | ||
| XM_006233363.2 | XP_006233425.1 | 6 | 1360 | |||
| XM_006233364.2 | XP_006233426.1 | 6 | 625 | |||
| GenBank mRNA | AY724517.1 | - | 6 | 994 | ||
| D90035.1 | - | 6 | 865 | |||
| FQ212355.1 | - | 6 | 1209 | |||
| M29275.1 | - | 6 | 865 | |||
| X57115.1 | - | 6 | 928 | |||

Ppp3ca
Genomic Map